Learn More
Medchemexpress LLC N-Arachidonyl maleimide | 876305-42-9 | 98.31% | 369.54 | 25 MG
SDP
Catalog No. 5000347940
Encompass_Preferred
N-Arachidonyl maleimide is a potent, irreversible inhibitor of monoacylglycerol lipase (MAGL) with an IC50 value of 140 nM. It can be used to modulate the effects of 2-arachidonyl glycerol (2-AG) by increasing its endogenous levels in the brain, and it prevents cerebellar membrane-mediated degradation of 2-AG.
  • Potent, irreversible MAGL inhibitor
  • Modulates effects of 2-arachidonyl glycerol (2-AG)
  • Increases endogenous levels of 2-AG in the brain
  • Prevents cerebellar membrane-mediated degradation of 2-AG
Catalog No. 50-003-47940 Supplier Medchemexpress LLC Supplier No. 25MGHY136562
N-Arachidonyl maleimide is a potent, irreversible inhibitor of monoacylglycerol lipase (MAGL) with an IC50 value of 140 nM. It can be used to modulate the effects of 2-arachidonyl glycerol (2-AG) by increasing its endogenous levels in the brain, and it prevents cerebellar membrane-mediated degradation of 2-AG.
  • Potent, irreversible MAGL inhibitor
  • Modulates effects of 2-arachidonyl glycerol (2-AG)
  • Increases endogenous levels of 2-AG in the brain
  • Prevents cerebellar membrane-mediated degradation of 2-AG