Learn More
Cayman Chemical N-ArachIdonyl TaurIn-d4 100ug
SDP
Catalog No. 502935380
Encompass_Preferred
An internal standard for the quantification of N-arachidonoyl taurine by GC- or LC-MS
Catalog No. 50-293-5380 Supplier Cayman Chemical Supplier No. 26323100 UG
Add to Cart
Add to Cart
An internal standard for the quantification of N-arachidonoyl taurine by GC- or LC-MS