Learn More
Cayman Chemical N-Arachidonoyl Taurine-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502935379
Encompass_Preferred
An internal standard for the quantification of N-arachidonoyl taurine by GC- or LC-MS
Catalog No. 50-293-5379 Supplier Cayman Chemical Supplier No. 263231 MG
An internal standard for the quantification of N-arachidonoyl taurine by GC- or LC-MS