Learn More
Cayman Chemical N-ArachIdonylAmInobutyrIc 10mg
SDP
Catalog No. 502895927
Encompass_Preferred
Several different arachidonoyl amino acids, including NAGABA, have been isolated and characterized from bovine brain. The glycine congener (NAGly) was found to suppress formalin-induced pain in rats. NAGABA also suppresses normal responses to pain, but has not yet been fully characterized.
Catalog No. 50-289-5927 Supplier Cayman Chemical Supplier No. 9006710 MG
Several different arachidonoyl amino acids, including NAGABA, have been isolated and characterized from bovine brain. The glycine congener (NAGly) was found to suppress formalin-induced pain in rats. NAGABA also suppresses normal responses to pain, but has not yet been fully characterized.