Learn More
Medchemexpress LLC N-Arachidoyl-D-sphingosine | 7344-02-7 | 99.1% | 594.01 | 50 MG
SDP
Catalog No. 5000439132
Encompass_Preferred
C20 Ceramide, also known as N-Arachidoyl-D-sphingosine, is a natural 20:0 ceramide. This N-acylsphingosine has its N-acyl group specified as eicosanoyl, functioning as a vital signal molecule.
  • Mediates cell growth
  • Influences cell differentiation
  • Involved in cell senescence
  • Regulates apoptosis
Catalog No. 50-004-39132 Supplier Medchemexpress LLC Supplier No. HY11201650MG
C20 Ceramide, also known as N-Arachidoyl-D-sphingosine, is a natural 20:0 ceramide. This N-acylsphingosine has its N-acyl group specified as eicosanoyl, functioning as a vital signal molecule.
  • Mediates cell growth
  • Influences cell differentiation
  • Involved in cell senescence
  • Regulates apoptosis