Learn More
Medchemexpress LLC N-(Azido-PEG2)-N-Boc-PEG3-Boc | 2093153-07-0 | C24H46N4O9 | 100 MG
SDP
Catalog No. 5000357039
Encompass_Preferred
N-(Azido-PEG2)-N-Boc-PEG3-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent with an azide group
  • Can be used in copper-catalyzed azide-alkyne cycloaddition reactions
  • Can be used in strain-promoted alkyne-azide cycloaddition reactions
  • Facilitates selective degradation of target proteins
Catalog No. 50-003-57039 Supplier Medchemexpress LLC Supplier No. HY140562100MG
N-(Azido-PEG2)-N-Boc-PEG3-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group that can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent with an azide group
  • Can be used in copper-catalyzed azide-alkyne cycloaddition reactions
  • Can be used in strain-promoted alkyne-azide cycloaddition reactions
  • Facilitates selective degradation of target proteins