Learn More
Medchemexpress LLC N-(Azido-PEG2)-N-Boc-PEG3-NHS ester | 2093153-85-4 | 97.0% | C24H41N5O11 | 250 MG
SDP
Catalog No. 5000352845
Encompass_Preferred
N-(Azido-PEG2)-N-Boc-PEG3-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a click chemistry reagent due to its Azide group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups.
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • For research use only.
Catalog No. 50-003-52845 Supplier Medchemexpress LLC Supplier No. 250MGHY140559
N-(Azido-PEG2)-N-Boc-PEG3-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a click chemistry reagent due to its Azide group.
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups.
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • For research use only.