Learn More
Medchemexpress LLC N-(Azido-PEG2)-N-Boc-PEG4-acid | 2093153-82-1 | C22H42N4O10 | 100 MG
SDP
Catalog No. 5000356904
Encompass_Preferred
N-(Azido-PEG2)-N-Boc-PEG4-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent with an Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-56904 Supplier Medchemexpress LLC Supplier No. HY140525100MG
N-(Azido-PEG2)-N-Boc-PEG4-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, which allows it to undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent with an Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions