Learn More
Medchemexpress LLC N-(Azido-PEG3)-N-Biotin-PEG4-methyl ester | 2100306-76-9 | 95.0% | C30H54N6O11S | 100 MG
SDP
Catalog No. 5000357020
Encompass_Preferred
N-(Azido-PEG3)-N-Biotin-PEG4-methyl ester is a PEG-based PROTAC linker for the synthesis of PROTACs. It also functions as a click chemistry reagent, facilitating reactions with various alkyne-containing, DBCO, or BCN molecules.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-57020 Supplier Medchemexpress LLC Supplier No. HY140581100MG
N-(Azido-PEG3)-N-Biotin-PEG4-methyl ester is a PEG-based PROTAC linker for the synthesis of PROTACs. It also functions as a click chemistry reagent, facilitating reactions with various alkyne-containing, DBCO, or BCN molecules.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC) reactions