Learn More
Medchemexpress LLC N-(Azido-PEG3)-N-Biotin-PEG4-methyl ester | 2100306-76-9 | 95.0% | C30H54N6O11S | 50 MG
SDP
Catalog No. 5000357055
Encompass_Preferred
N-(Azido-PEG3)-N-Biotin-PEG4-methyl ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, enabling specific reactions with other molecules. This product is intended for research use only.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group for specific reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-57055 Supplier Medchemexpress LLC Supplier No. HY14058150MG
N-(Azido-PEG3)-N-Biotin-PEG4-methyl ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, enabling specific reactions with other molecules. This product is intended for research use only.
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group for specific reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions