Learn More
Medchemexpress LLC N-(Azido-PEG3)-N-(PEG2-NH-Boc)-PEG3-acid | 2183440-74-4 | 98.1% | C29H55N5O13 | 10MG
SDP
Catalog No. 5000259308
Encompass_Preferred
N-(Azido-PEG3)-N-(PEG2-NH-Boc)-PEG3-acid is a PEG-based linker designed for use in click chemistry and bioconjugation workflows. It features a terminal azide and orthogonally protected amine, providing versatility for PROTAC assembly and other synthetic applications.
  • Terminal azide for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC).
  • PEG3 spacer provides flexibility and improved solubility.
  • Boc-protected amine on PEG2 enables selective deprotection and further modification.
  • Carboxylic acid terminus allows conjugation via standard amide coupling.
  • High purity suitable for synthetic and bioconjugation workflows.
Catalog No. 50-002-59308 Supplier Medchemexpress LLC Supplier No. HY14128710MG
N-(Azido-PEG3)-N-(PEG2-NH-Boc)-PEG3-acid is a PEG-based linker designed for use in click chemistry and bioconjugation workflows. It features a terminal azide and orthogonally protected amine, providing versatility for PROTAC assembly and other synthetic applications.
  • Terminal azide for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Compatible with strain-promoted alkyne-azide cycloaddition (SPAAC).
  • PEG3 spacer provides flexibility and improved solubility.
  • Boc-protected amine on PEG2 enables selective deprotection and further modification.
  • Carboxylic acid terminus allows conjugation via standard amide coupling.
  • High purity suitable for synthetic and bioconjugation workflows.