Learn More
Medchemexpress LLC N-(Azido-PEG4)-N-bis(PEG4-acid) | 2093152-80-6 | 100 MG
SDP
Catalog No. 5000356229
Encompass_Preferred
N-(Azido-PEG4)-N-bis(PEG4-acid) is a PROTAC linker belonging to the PEG class, used for synthesizing PROTAC molecules. It is a click chemistry reagent containing an Azide group, which can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • PROTAC linker belonging to the PEG class
  • Used for synthesizing PROTAC molecules
  • Click chemistry reagent containing an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-56229 Supplier Medchemexpress LLC Supplier No. HY140519100MG
N-(Azido-PEG4)-N-bis(PEG4-acid) is a PROTAC linker belonging to the PEG class, used for synthesizing PROTAC molecules. It is a click chemistry reagent containing an Azide group, which can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • PROTAC linker belonging to the PEG class
  • Used for synthesizing PROTAC molecules
  • Click chemistry reagent containing an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups
  • Utilizes the intracellular ubiquitin-proteasome system to selectively degrade target proteins