Learn More
Medchemexpress LLC N-(Azido-PEG4)-N-Boc-PEG4-NHS ester | 2112731-96-9 | ≥97.0% | 25 MG
SDP
Catalog No. 5000341432
Encompass_Preferred
N-(Azido-PEG4)-N-Boc-PEG4-NHS ester is a PEG-based PROTAC linker, primarily utilized in the synthesis of PROTACs. It functions as a versatile click chemistry reagent due to its terminal azide group. This enables it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN group-containing molecules.
  • PEG-based PROTAC linker with a terminal azide group
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
Catalog No. 50-003-41432 Supplier Medchemexpress LLC Supplier No. 25MGHY130312
N-(Azido-PEG4)-N-Boc-PEG4-NHS ester is a PEG-based PROTAC linker, primarily utilized in the synthesis of PROTACs. It functions as a versatile click chemistry reagent due to its terminal azide group. This enables it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN group-containing molecules.
  • PEG-based PROTAC linker with a terminal azide group
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups