Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Azidoacetylgalactosamine-Tetraacylated

Catalog No. 90091550MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-091-550MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-091-550MG Supplier Sigma Aldrich Supplier No. 90091550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Azidoacetylgalactosamine-tetraacylated (Ac4GalNAz) provides the first part of a simple and robust two-step technique that helps identify and characterize cell surface sialic acid-containing glycoproteins. The azide-modified protein can be detected by reaction with alkynes. For example alkynes labeled with a fluorescent probe or a biotin can be used. The acetyl groups increase cell permeability and allow the unnatural sugars to easily pass through the cell membrane. Carboxyesterases remove the acetyl groups once the monosaccharide is in the cell.

Specifications

CAS 365564-05-2
Molecular Formula C15H13N5O4
Linear Formula C15H13N5O4
MDL Number MFCD28334557
Quantity 100 mg
Molecular Weight (g/mol) 327.29 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.