Learn More
Enzo Life Sciences N-Butyldeoxynojirimycin. HCl (25 mg). CAS: 210110-90-0
SDP
Supplier:  Enzo Life Sciences BMLSL2300025
Encompass_Preferred
A novel inhibitor of glucosylceramide synthase as well as glucosidase I and II. Inhibits glucosylceramide synthase without ceramide accumulation and cell death. It is a potent inhibitor of human immunodeficiency virus (HIV) replication and syncytium formation in vitro. Restores chloride current in epithelial CF cells. Alternative name: Miglustat. Purity: ≥98% (TLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-2139