Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Behenoyl-D-erythro-sphingosine | 27888-44-4 | MFCD09840213 | 98.0% | 622.06 g/mol | C40H79NO3 | 10 MG
SDP

Catalog No. 5000285955
Encompass_Preferred

C22-Ceramide (N-Behenoyl-D-erythro-sphingosine) is an endogenous bioactive sphingolipid used in research on membrane dynamics and mitochondrial ceramide channel formation. It reduces the propensity of C16-ceramide channel formation in isolated rat liver mitochondria and in liposomes. Supplied as a solid white to off-white powder with high purity for analytical and biological studies.

  • Endogenous bioactive sphingolipid relevant to membrane and apoptosis studies.
  • Reduces propensity of C16-ceramide channel formation in mitochondria and liposomes.
  • White to off-white solid suitable for handling and formulation.
  • Purity 98.0% for reproducible experimental results.
  • Molecular formula C40H79NO3; molecular weight 622.06 g/mol.
  • Soluble in ethanol and DMF using ultrasonic and warming up to 60°C as needed.
  • Store at 4°C protected from light; in solution keep at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-85955 Supplier Medchemexpress LLC Supplier No. HY15483110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C22-Ceramide (N-Behenoyl-D-erythro-sphingosine) is an endogenous bioactive sphingolipid used in research on membrane dynamics and mitochondrial ceramide channel formation. It reduces the propensity of C16-ceramide channel formation in isolated rat liver mitochondria and in liposomes. Supplied as a solid white to off-white powder with high purity for analytical and biological studies.

  • Endogenous bioactive sphingolipid relevant to membrane and apoptosis studies.
  • Reduces propensity of C16-ceramide channel formation in mitochondria and liposomes.
  • White to off-white solid suitable for handling and formulation.
  • Purity 98.0% for reproducible experimental results.
  • Molecular formula C40H79NO3; molecular weight 622.06 g/mol.
  • Soluble in ethanol and DMF using ultrasonic and warming up to 60°C as needed.
  • Store at 4°C protected from light; in solution keep at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.