Learn More
Medchemexpress LLC N'-(benzenesulfonyl)-2-fluoro-5-(pyridazin-4-yl)benzohydrazide | 2055397-88-9 | 99.6% | 372.37 | C17H13FN4O3S | 5 MG
SDP
Catalog No. 5000245713
Encompass_Preferred
MOZ-IN-2 is a small-molecule inhibitor of the histone acetyltransferase MOZ (KAT6A) with a reported IC50 of 125 μM. Provided as an off-white to light yellow solid powder, it is intended for research use in biochemical assays and epigenetic studies investigating MOZ function and modulation.
  • Inhibits MOZ histone acetyltransferase (IC50 125 μM).
  • High purity (~99.6%) suitable for research assays.
  • Off-white to light yellow solid form for convenient handling and storage.
  • Well-characterized molecular properties (MW 372.37; formula C17H13FN4O3S).
  • Stable under recommended storage conditions for solid samples.
Catalog No. 50-002-45713 Supplier Medchemexpress LLC Supplier No. HY1020595MG
MOZ-IN-2 is a small-molecule inhibitor of the histone acetyltransferase MOZ (KAT6A) with a reported IC50 of 125 μM. Provided as an off-white to light yellow solid powder, it is intended for research use in biochemical assays and epigenetic studies investigating MOZ function and modulation.
  • Inhibits MOZ histone acetyltransferase (IC50 125 μM).
  • High purity (~99.6%) suitable for research assays.
  • Off-white to light yellow solid form for convenient handling and storage.
  • Well-characterized molecular properties (MW 372.37; formula C17H13FN4O3S).
  • Stable under recommended storage conditions for solid samples.