Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N'-(benzenesulfonyl)-2-fluoro-5-pyridazin-4-ylbenzohydrazide | 2055397-88-9 | MFCD31617713 | 99.6% | 372.37 g/mol | C17H13FN4O3S | 10 MG
SDP

Catalog No. 5000245601
Encompass_Preferred

MOZ-IN-2 is a small-molecule inhibitor of MOZ (KAT6A) histone acetyltransferase with a reported IC50 of 125 μM. It is supplied as a solid powder and as a 10 mM DMSO solution for research use and is characterized by high purity and defined molecular properties.

  • Inhibits MOZ (KAT6A) with IC50 = 125 μM.
  • Available as solid (5 mg, 10 mg, 25 mg) and 10 mM solution in DMSO.
  • Purity approximately 99.6%.
  • Molecular weight 372.37 g/mol; formula C17H13FN4O3S.
  • Suitable for biochemical and cell-based assays.
  • Provided with Safety Data Sheet and characterization data.

Catalog No. 50-002-45601 Supplier Medchemexpress LLC Supplier No. HY10205910MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MOZ-IN-2 is a small-molecule inhibitor of MOZ (KAT6A) histone acetyltransferase with a reported IC50 of 125 μM. It is supplied as a solid powder and as a 10 mM DMSO solution for research use and is characterized by high purity and defined molecular properties.

  • Inhibits MOZ (KAT6A) with IC50 = 125 μM.
  • Available as solid (5 mg, 10 mg, 25 mg) and 10 mM solution in DMSO.
  • Purity approximately 99.6%.
  • Molecular weight 372.37 g/mol; formula C17H13FN4O3S.
  • Suitable for biochemical and cell-based assays.
  • Provided with Safety Data Sheet and characterization data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.