Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Benzoyl-(2R,3S)-3-phenylisoserine | 132201-33-3 | 25 MG
SDP

Catalog No. 5000420621
Encompass_Preferred

N-Benzoyl-(2R,3S)-3-phenylisoserine is a Taxol C-13 Side Chain, crucial for the strong antitumor activity of Taxol. It has a molecular weight of 285.29 and a chemical formula of C16H15NO4. This compound appears as a white to off-white solid with a purity of 99.48%.

  • Crucial for the strong antitumor activity of Taxol
  • Taxol C-13 side chain
  • Purity of 99.48%
  • Molecular weight: 285.29
  • Chemical formula: C16H15NO4
  • Appears as a white to off-white solid
  • Structure classification: Alkaloids, other alkaloids
  • Initial source: Plants, Taxaceae, *Taxus wallichiana Zucc.*

Catalog No. 50-004-20621 Supplier Medchemexpress LLC Supplier No. 25MGHYN2380
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Benzoyl-(2R,3S)-3-phenylisoserine is a Taxol C-13 Side Chain, crucial for the strong antitumor activity of Taxol. It has a molecular weight of 285.29 and a chemical formula of C16H15NO4. This compound appears as a white to off-white solid with a purity of 99.48%.

  • Crucial for the strong antitumor activity of Taxol
  • Taxol C-13 side chain
  • Purity of 99.48%
  • Molecular weight: 285.29
  • Chemical formula: C16H15NO4
  • Appears as a white to off-white solid
  • Structure classification: Alkaloids, other alkaloids
  • Initial source: Plants, Taxaceae, *Taxus wallichiana Zucc.*

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.