Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Benzoyl-(2R,3S)-3-phenylisoserine | 132201-33-3 | 5 MG
SDP

Catalog No. 5000421321
Encompass_Preferred

N-Benzoyl-(2R,3S)-3-phenylisoserine is a Taxol C-13 Side Chain that is crucial for the strong antitumor activity of Taxol.

  • Purity of 99.48%
  • Appearance as a white to off-white solid
  • Soluble in DMSO at ≥ 160 mg/mL (560.83 mM)

Catalog No. 50-004-21321 Supplier Medchemexpress LLC Supplier No. HYN23805MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Benzoyl-(2R,3S)-3-phenylisoserine is a Taxol C-13 Side Chain that is crucial for the strong antitumor activity of Taxol.

  • Purity of 99.48%
  • Appearance as a white to off-white solid
  • Soluble in DMSO at ≥ 160 mg/mL (560.83 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.