Learn More
Medchemexpress LLC N-Benzoyl-(2R,3S)-3-phenylisoserine | 132201-33-3 | 50 MG
SDP
Catalog No. 5000420676
Encompass_Preferred
N-Benzoyl-(2R,3S)-3-phenylisoserine is a Taxol C-13 Side Chain, crucial for the strong antitumor activity of Taxol. It has a molecular weight of 285.29, a chemical formula of C16H15NO4, and appears as a white to off-white solid with 99.48% purity.
  • Crucial for the strong antitumor activity of Taxol
  • Exhibits cellular effects, showing 62% inhibition in B16 melanoma cells at 42 μM
  • Has an IC50 of > 100 μM in rat L6 cells after 70 hours
  • White to off-white solid appearance
  • Purity of 99.48%
Catalog No. 50-004-20676 Supplier Medchemexpress LLC Supplier No. HYN238050MG
N-Benzoyl-(2R,3S)-3-phenylisoserine is a Taxol C-13 Side Chain, crucial for the strong antitumor activity of Taxol. It has a molecular weight of 285.29, a chemical formula of C16H15NO4, and appears as a white to off-white solid with 99.48% purity.
  • Crucial for the strong antitumor activity of Taxol
  • Exhibits cellular effects, showing 62% inhibition in B16 melanoma cells at 42 μM
  • Has an IC50 of > 100 μM in rat L6 cells after 70 hours
  • White to off-white solid appearance
  • Purity of 99.48%