Learn More
Medchemexpress LLC N-Benzoyl-β-alanine | 3440-28-6 | 98.7% | 10 G
SDP
Catalog No. 5000413491
Encompass_Preferred
Betamipron is a chemical compound primarily used in conjunction with Panipenem. It functions by inhibiting Panipenem uptake into the renal tubule, thereby preventing nephrotoxicity. This compound is suitable for various laboratory applications and for the manufacture of substances.
  • Inhibits Panipenem uptake in the renal tubule.
  • Prevents nephrotoxicity.
  • Shows an IC50 of 6 μM for inhibiting Adefovir uptake in OAT1-expressing CHO cells.
  • Soluble in DMSO at 100 mg/mL.
  • Soluble in various in vivo protocols at concentrations ≥ 2.5 mg/mL.
Catalog No. 50-004-13491 Supplier Medchemexpress LLC Supplier No. HYB112710G
Betamipron is a chemical compound primarily used in conjunction with Panipenem. It functions by inhibiting Panipenem uptake into the renal tubule, thereby preventing nephrotoxicity. This compound is suitable for various laboratory applications and for the manufacture of substances.
  • Inhibits Panipenem uptake in the renal tubule.
  • Prevents nephrotoxicity.
  • Shows an IC50 of 6 μM for inhibiting Adefovir uptake in OAT1-expressing CHO cells.
  • Soluble in DMSO at 100 mg/mL.
  • Soluble in various in vivo protocols at concentrations ≥ 2.5 mg/mL.