Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nα-Benzoyl-L-arginine 4-nitroanilide (hydrochloride) | 21653-40-7 | 99.9% | 434.88 | 1 ML
SDP

Catalog No. 5000351829
Encompass_Preferred

Nα-Benzoyl-L-arginine 4-nitroanilide (hydrochloride) is an arginine derivative primarily used in various research applications. It is supplied as a solution and typically appears as a white to light yellow solid.

  • Arginine derivative.
  • Features solid appearance, with color ranging from white to light yellow.
  • Molecular weight of 434.88.
  • Purity of 99.9%.
  • CAS Number is 21653-40-7.
  • Solid storage at 4°C, sealed, away from moisture.
  • Solution storage at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture.

Catalog No. 50-003-51829 Supplier Medchemexpress LLC Supplier No. HY13752910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nα-Benzoyl-L-arginine 4-nitroanilide (hydrochloride) is an arginine derivative primarily used in various research applications. It is supplied as a solution and typically appears as a white to light yellow solid.

  • Arginine derivative.
  • Features solid appearance, with color ranging from white to light yellow.
  • Molecular weight of 434.88.
  • Purity of 99.9%.
  • CAS Number is 21653-40-7.
  • Solid storage at 4°C, sealed, away from moisture.
  • Solution storage at -80°C for 6 months or -20°C for 1 month, sealed, away from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.