Learn More
Medchemexpress LLC N-benzoyl-L-aspartic acid | 997815 | MFCD01664832 | 237.21 g/mol | C11H11NO5 | 500 MG
SDP
Catalog No. 5000227296
Encompass_Preferred
N-benzoyl-L-aspartic acid is an analytical reference standard of a major metabolite of benzyl glucosinolate supplied for research and analytical applications. It is intended for use in method development, qualitative and quantitative assays, and peptide or protein modification.
  • Analytical reference standard for assay development and validation.
  • Compatible with HPLC, GC, and MS workflows.
  • Can be used for modification of peptides or proteins.
  • Molecular formula C11H11NO5; molecular weight 237.21 g/mol.
  • Available in multiple pack sizes, including 500 MG.
Catalog No. 50-002-27296 Supplier Medchemexpress LLC Supplier No. HY118093500MG
N-benzoyl-L-aspartic acid is an analytical reference standard of a major metabolite of benzyl glucosinolate supplied for research and analytical applications. It is intended for use in method development, qualitative and quantitative assays, and peptide or protein modification.
  • Analytical reference standard for assay development and validation.
  • Compatible with HPLC, GC, and MS workflows.
  • Can be used for modification of peptides or proteins.
  • Molecular formula C11H11NO5; molecular weight 237.21 g/mol.
  • Available in multiple pack sizes, including 500 MG.