Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzoyl-L-glutamic acid, 98%, Thermo Scientific™

Catalog No. AAH6677814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6677814 25 g
AAH6677806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6677814 Supplier Thermo Scientific Chemicals Supplier No. H6677814

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6094-36-6
Molecular Formula C12H13NO5
Molecular Weight (g/mol) 251.238
MDL Number MFCD00014028
InChI Key LPJXPACOXRZCCP-VIFPVBQESA-N
Synonym bz-glu-oh, n-benzoyl-l-glutamic acid, benzoyl-l-glutamic acid, s-2-benzamidopentanedioic acid, n-benzoylglutamic acid, unii-3591d74mxp, 2s-2-phenylformamido pentanedioic acid, +-n-benzoylglutamic acid, n-4-benzoyl-l-glutamic acid, 2s-2-phenylcarbonylamino pentanedioic acid
PubChem CID 11345724
IUPAC Name (2S)-2-benzamidopentanedioic acid
SMILES C1=CC=C(C=C1)C(=O)NC(CCC(=O)O)C(=O)O

Specifications

Beilstein 2622989
Quantity 25 g
Formula Weight 251.24
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Benzoyl-L-glutamic acid
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.