Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-benzoyl-L-leucine | 1466-83-7 | MFCD00190766 | 99.6% | 235.28 g·mol⁻¹ | C13H17NO3 | 5 G
SDP

Catalog No. 5000282985
Encompass_Preferred

N-benzoyl-L-leucine is an N-benzoylated derivative of the amino acid L-leucine used as a research reagent and reference standard in biochemical and pharmacological studies. It is supplied as a white to off-white solid, with molecular formula C13H17NO3 and molecular weight 235.28 g·mol⁻¹. Typical solubility in dimethyl sulfoxide (DMSO) is high, facilitating solution preparation for assays.

  • White to off-white solid, stable powder
  • Molecular formula C13H17NO3; molecular weight 235.28 g·mol⁻¹
  • Purity reported at approximately 99.6%
  • Soluble in DMSO (250 mg/mL) with ultrasonic assistance
  • Storage conditions support multiyear stability for the powder
  • Available in research quantities such as 5 G

Catalog No. 50-002-82985 Supplier Medchemexpress LLC Supplier No. HYW1420355G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-benzoyl-L-leucine is an N-benzoylated derivative of the amino acid L-leucine used as a research reagent and reference standard in biochemical and pharmacological studies. It is supplied as a white to off-white solid, with molecular formula C13H17NO3 and molecular weight 235.28 g·mol⁻¹. Typical solubility in dimethyl sulfoxide (DMSO) is high, facilitating solution preparation for assays.

  • White to off-white solid, stable powder
  • Molecular formula C13H17NO3; molecular weight 235.28 g·mol⁻¹
  • Purity reported at approximately 99.6%
  • Soluble in DMSO (250 mg/mL) with ultrasonic assistance
  • Storage conditions support multiyear stability for the powder
  • Available in research quantities such as 5 G

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.