Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzoyl-L-tyrosine 98.0+%, TCI America™
SDP

Catalog No. B19115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19115G 5 g
B191125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19115G Supplier TCI America Supplier No. B19115G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2566-23-6
Molecular Formula C16H15NO4
Molecular Weight (g/mol) 285.299
MDL Number MFCD01321140
InChI Key KUUUDPTUEOKITK-AWEZNQCLSA-N
PubChem CID 151021
IUPAC Name (2S)-2-benzamido-3-(4-hydroxyphenyl)propanoic acid
SMILES C1=CC=C(C=C1)C(=O)NC(CC2=CC=C(C=C2)O)C(=O)O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 285.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzoyl-L-tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.