Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzoylaminopurine, 99%

Catalog No. p-7044802
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0829203 1 g
AAL0829206 5 g
AAL0829214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0829203 Supplier Thermo Scientific Chemicals Supplier No. L0829203
Only null left
Add to Cart

CAS: 4005-49-6 | C12H9N5O | 239.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4005-49-6
Molecular Formula C12H9N5O
Molecular Weight (g/mol) 239.238
MDL Number MFCD00037927
InChI Key QQJXZVKXNSFHRI-UHFFFAOYSA-N
Synonym n6-benzoyladenine, n-7h-purin-6-yl benzamide, n-benzoylaminopurine, n-9h-purin-6-yl benzamide, 6-benzoylaminopurine, benzamide, n-1h-purin-6-yl, 6-benzamidopurine, adenine,6n-benzoyl, phenyl-n-purin-6-ylcarboxamide, n-benzoyladenine
PubChem CID 97075
IUPAC Name N-(7H-purin-6-yl)benzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=NC=NC3=C2NC=N3

Specifications

Melting Point 241°C to 244°C
Quantity 1 g
Beilstein 20585
Formula Weight 239.23
Percent Purity 99%
Chemical Name or Material N-Benzoylaminopurine
Hazard Category H302-H317
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.