Learn More
Medchemexpress LLC N-Benzoylcytidine | 13089-48-0 | 99.5% | C16H17N3O6 | 1 ML
SDP
Catalog No. 5000433289
Encompass_Preferred
N-Benzoylcytidine is a modified cytidine analogue that can be phosphorylated by uracil-cytidine kinase (UCK1 and UCK2). It can be used to synthesize 2-OH protective groups for solid-phase RNA synthesis and synthetic oligonucleotides for UV induction and targeted gene silencing in zebrafish embryos.
  • Modified cytidine analogue
  • Can be phosphorylated by uracil-cytidine kinase (UCK1 and UCK2)
  • Useful for synthesizing 2-OH protective groups for solid-phase RNA synthesis
  • Can be used to synthesize oligonucleotides for UV induction and targeted gene silencing in zebrafish embryos
  • For research use only
Catalog No. 50-004-33289 Supplier Medchemexpress LLC Supplier No. HY7857410MM/1ML
N-Benzoylcytidine is a modified cytidine analogue that can be phosphorylated by uracil-cytidine kinase (UCK1 and UCK2). It can be used to synthesize 2-OH protective groups for solid-phase RNA synthesis and synthetic oligonucleotides for UV induction and targeted gene silencing in zebrafish embryos.
  • Modified cytidine analogue
  • Can be phosphorylated by uracil-cytidine kinase (UCK1 and UCK2)
  • Useful for synthesizing 2-OH protective groups for solid-phase RNA synthesis
  • Can be used to synthesize oligonucleotides for UV induction and targeted gene silencing in zebrafish embryos
  • For research use only