Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-4-(chloromethyl)-N-isopropylbenzamide, 97%, Thermo Scientific™

Catalog No. AAH6018403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6018403 1 g
AAH60184MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6018403 Supplier Thermo Scientific Chemicals Supplier No. H6018403

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1097079-85-0
Molecular Formula C18H20ClNO
Molecular Weight (g/mol) 301.814
MDL Number MFCD13658458
InChI Key BLONTFYZTPFISN-UHFFFAOYSA-N
Synonym n-benzyl-4-chloromethyl-n-isopropylbenzamide
PubChem CID 43333466
IUPAC Name N-benzyl-4-(chloromethyl)-N-propan-2-ylbenzamide
SMILES CC(C)N(CC1=CC=CC=C1)C(=O)C2=CC=C(C=C2)CCl

Specifications

Quantity 1 g
Formula Weight 301.81
Percent Purity 97%
Chemical Name or Material N-Benzyl-4-(chloromethyl)-N-isopropylbenzamide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.