Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-4-methoxy-N-phenylbenzamide, 97%, Thermo Scientific™

Catalog No. AAH59124MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH59124MD 250 mg
AAH5912403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH59124MD Supplier Thermo Scientific Chemicals Supplier No. H59124MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111735-27-4
Molecular Formula C21H19NO2
Molecular Weight (g/mol) 317.39
MDL Number MFCD22683043
InChI Key BLUMCNOTEFEZGV-UHFFFAOYSA-N
Synonym benzamide, 4-methoxy-n-phenyl-n-phenylmethyl, acmc-20mep6
PubChem CID 13736433
IUPAC Name N-benzyl-4-methoxy-N-phenylbenzamide
SMILES COC1=CC=C(C=C1)C(=O)N(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Quantity 250 mg
Formula Weight 317.39
Percent Purity 97%
Chemical Name or Material N-Benzyl-4-methoxy-N-phenylbenzamide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.