Learn More
Medchemexpress LLC N-benzyl-4-(N-benzylsulfamoyl)benzamide | 313520-94-4 | MFCD00990583 | 99.0% | C21H20N2O3S | 10MG
SDP
Catalog No. 5000261348
Encompass_Preferred
TH-263 is a diaryl sulfonamide derivative (CAS 313520-94-4) that is inactive toward both LIMK1 and LIMK2 and is used as a negative control for the LIMK inhibitor TH-257. It is supplied as a high-purity solid (molecular formula C21H20N2O3S, MW 380.46 g/mol), soluble in DMSO, and intended for use as a selectivity control in biochemical and cell-based assays.
  • Diaryl sulfonamide derivative inactive toward LIMK1 and LIMK2.
  • High purity: 99.0%.
  • Suitable as a negative control in biochemical and cell-based assays.
  • Soluble in DMSO for in vitro applications.
  • Powder stable at -20°C; solutions stored at -80°C for extended stability.
Catalog No. 50-002-61348 Supplier Medchemexpress LLC Supplier No. HY13584310MG
TH-263 is a diaryl sulfonamide derivative (CAS 313520-94-4) that is inactive toward both LIMK1 and LIMK2 and is used as a negative control for the LIMK inhibitor TH-257. It is supplied as a high-purity solid (molecular formula C21H20N2O3S, MW 380.46 g/mol), soluble in DMSO, and intended for use as a selectivity control in biochemical and cell-based assays.
  • Diaryl sulfonamide derivative inactive toward LIMK1 and LIMK2.
  • High purity: 99.0%.
  • Suitable as a negative control in biochemical and cell-based assays.
  • Soluble in DMSO for in vitro applications.
  • Powder stable at -20°C; solutions stored at -80°C for extended stability.