Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Benzyl-L-Proline Ethyl Ester

Catalog No. 3692925G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-929-25G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 36-929-25G Supplier Sigma Aldrich Supplier No. 3692925G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Benzyl-L-Proline Ethyl Ester

Specifications

CAS 17442-18-1
Melting Point 205°C
Molecular Formula C36H30Cl3OP2Re
Linear Formula [(C6H5)3P]2ReOCl3
Refractive Index n20/D 1.511
MDL Number MFCD00075554
Quantity 1 g
Synonym Bis(triphenylphosphine)rhenium oxytrichloride; Bis(triphenylphosphine)trichlororhenium oxide
Molecular Weight (g/mol) 833.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.