Learn More
Cayman Chemical N-Benzyl LInoleamIde 10mg
SDP
Supplier:  Cayman Chemical 3335810 MG
Encompass_Preferred
A soluble epoxide hydrolase inhibitor (IC50s = 0.155, 0.041, and 0.044 µM for the human, rat, and mouse enzymes, respectively); selective for sEH over FAAH (IC50 = 10.8 µM for the human enzyme); inhibits production of NF-κB (IC50 = 8.80 µM) and activates Nrf2 (EC50 = 35.24 µM) in Neuro2a cells; increases the paw withdrawal threshold in a mouse model of LPS-induced inflammatory pain at 100 mg/kg
Catalog No. 50-294-9946