Learn More
Medchemexpress LLC N-benzyl-N-butyl-4-(phenylsulfamoyl)benzamide | 2244678-29-1 | 98.4% | 50 MG
SDP
Catalog No. 5000230254
Encompass_Preferred
TH-257 (CAS 2244678-29-1) is a small-molecule allosteric inhibitor of LIM kinase 1 and 2 used as a biochemical research probe to study LIMK-mediated signaling pathways in vitro.
  • Potent inhibitor of LIMK1 and LIMK2 with IC50 values of 84 nM and 39 nM.
  • Molecular formula C24H26N2O3S and molecular weight 422.54 g/mol.
  • High purity (98.36%) suitable for in vitro biochemical studies.
  • Supplied as a solid powder and as a 10 mM solution in DMSO; common pack sizes include 5 mg, 10 mg, 50 mg, and 100 mg.
Catalog No. 50-002-30254 Supplier Medchemexpress LLC Supplier No. HY12263050MG
TH-257 (CAS 2244678-29-1) is a small-molecule allosteric inhibitor of LIM kinase 1 and 2 used as a biochemical research probe to study LIMK-mediated signaling pathways in vitro.
  • Potent inhibitor of LIMK1 and LIMK2 with IC50 values of 84 nM and 39 nM.
  • Molecular formula C24H26N2O3S and molecular weight 422.54 g/mol.
  • High purity (98.36%) suitable for in vitro biochemical studies.
  • Supplied as a solid powder and as a 10 mM solution in DMSO; common pack sizes include 5 mg, 10 mg, 50 mg, and 100 mg.