Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-N-ethyl-4-nitrobenzamide, 97%, Thermo Scientific™

Catalog No. AAH60983MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60983MD 250 mg
AAH6098303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60983MD Supplier Thermo Scientific Chemicals Supplier No. H60983MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349396-05-0
Molecular Formula C16H16N2O3
Molecular Weight (g/mol) 284.32
MDL Number MFCD01212056
InChI Key MWXUHJACDKHYRK-UHFFFAOYSA-N
Synonym cbmicro_009552, n-ethyl 4-nitrophenyl-n-benzylcarboxamide
PubChem CID 779809
IUPAC Name N-benzyl-N-ethyl-4-nitrobenzamide
SMILES CCN(CC1=CC=CC=C1)C(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Quantity 250 mg
Formula Weight 284.31
Percent Purity 97%
Chemical Name or Material N-Benzyl-N-ethyl-4-nitrobenzamide
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.