Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-N-ethylaniline 98.0+%, TCI America™
SDP

Catalog No. E0162500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0162500G 500 g
E016225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0162500G Supplier TCI America Supplier No. E0162500G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 92-59-1
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.31
MDL Number MFCD00009037
InChI Key HSZCJVZRHXPCIA-UHFFFAOYSA-N
Synonym ethylbenzylaniline, benzenemethanamine, n-ethyl-n-phenyl, n-ethyl-n-phenylbenzylamine, benzylethylphenylamine, phenylethylbenzylamine, n-ethyl-n-benzylaniline, benzylamine, n-ethyl-n-phenyl, n-ethyl-n-phenyl benzylamine, n-ethyl-n-phenylbenzenemethanamine, amine, benzyl ethyl phenyl
PubChem CID 7098
IUPAC Name N-benzyl-N-ethylaniline
SMILES CCN(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 287°C
Quantity 500 g
UN Number 2274
Formula Weight 211.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Benzyl-N-ethylaniline
EINECSNumber (3)-0377
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.