Learn More
Medchemexpress LLC N-benzyl-N-hydroxy-3,3-dimethylbutanamide | 1956370-21-0 | MFCD30738006 | 99.9% | 221.30 g/mol | C13H19NO2 | 50 MG
SDP
Catalog No. 5000213567
Encompass_Preferred
RIPA-56 is a potent, selective, and metabolically stable inhibitor of receptor-interacting protein 1 (RIP1) with an IC50 of 13 nM. Supplied as a solid 50 mg quantity, it has the chemical formula C13H19NO2 and a molecular weight of approximately 221.3 g/mol. The compound is supplied at high purity for research use and includes recommended storage conditions for powder and solutions.
Catalog No. 50-002-13567 Supplier Medchemexpress LLC Supplier No. HY10103250MG
RIPA-56 is a potent, selective, and metabolically stable inhibitor of receptor-interacting protein 1 (RIP1) with an IC50 of 13 nM. Supplied as a solid 50 mg quantity, it has the chemical formula C13H19NO2 and a molecular weight of approximately 221.3 g/mol. The compound is supplied at high purity for research use and includes recommended storage conditions for powder and solutions.