Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylacetoacetamide 98.0+%, TCI America™
SDP

Catalog No. B280825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B280825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B280825G Supplier TCI America Supplier No. B280825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 882-36-0
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD00026260
InChI Key KOHNUEXAOQRRPI-UHFFFAOYSA-N
Synonym N-Acetoacetylbenzylamine
PubChem CID 70159
IUPAC Name N-benzyl-3-oxobutanamide
SMILES CC(=O)CC(=O)NCC1=CC=CC=C1

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 25 g
Formula Weight 191.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzylacetoacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.