Learn More
Medchemexpress LLC N-benzylaniline | 103-32-2 | MFCD00003018 | 99.9% | 183.25 | C13H13N | 1 ML
SDP
Catalog No. 5000303721
Encompass_Preferred
N-Benzylaniline (N-Phenylbenzylamine) is an N-alkylated derivative of aniline supplied as a 10 mM solution in DMSO. It is intended for use as a reagent or reference standard in chemical, biochemical, and analytical workflows and is characterized by its stated purity and molecular properties (CAS 103-32-2; C13H13N; MW 183.25).
  • Ready-to-use 10 mM solution in DMSO, 1 mL volume.
  • High purity (99.9%) appropriate for analytical applications.
  • Convenient small-volume format reduces freeze-thaw and contamination.
  • Clear physicochemical description: light yellow to brown; solid below 35°C, liquid above 38°C.
  • Supplied with datasheet and batch information for traceability.
Catalog No. 50-003-03721 Supplier Medchemexpress LLC Supplier No. HYW01632110MM/1ML
N-Benzylaniline (N-Phenylbenzylamine) is an N-alkylated derivative of aniline supplied as a 10 mM solution in DMSO. It is intended for use as a reagent or reference standard in chemical, biochemical, and analytical workflows and is characterized by its stated purity and molecular properties (CAS 103-32-2; C13H13N; MW 183.25).
  • Ready-to-use 10 mM solution in DMSO, 1 mL volume.
  • High purity (99.9%) appropriate for analytical applications.
  • Convenient small-volume format reduces freeze-thaw and contamination.
  • Clear physicochemical description: light yellow to brown; solid below 35°C, liquid above 38°C.
  • Supplied with datasheet and batch information for traceability.