Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-benzylaniline (N-phenylbenzylamine) | 103-32-2 | MFCD00003018 | 99.9% | 183.25 g/mol | C13H13N | 10 G
SDP

Catalog No. 5000302192
Encompass_Preferred

N-Benzylaniline (N-Phenylbenzylamine) is an N-alkylated derivative of aniline offered as a high-purity research chemical. It is used as a building block and reference standard in organic synthesis and medicinal chemistry, and is typically handled neat or prepared as DMSO stock solutions.

  • High purity suitable for research applications.
  • Useful building block for organic synthesis.
  • Suitable as a reference standard in analytical methods.
  • Solid or liquid depending on temperature; light yellow to brown appearance.
  • Molecular formula C13H13N; molecular weight 183.25 g/mol.
  • Soluble in DMSO for preparation of stock solutions.

Catalog No. 50-003-02192 Supplier Medchemexpress LLC Supplier No. HYW01632110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Benzylaniline (N-Phenylbenzylamine) is an N-alkylated derivative of aniline offered as a high-purity research chemical. It is used as a building block and reference standard in organic synthesis and medicinal chemistry, and is typically handled neat or prepared as DMSO stock solutions.

  • High purity suitable for research applications.
  • Useful building block for organic synthesis.
  • Suitable as a reference standard in analytical methods.
  • Solid or liquid depending on temperature; light yellow to brown appearance.
  • Molecular formula C13H13N; molecular weight 183.25 g/mol.
  • Soluble in DMSO for preparation of stock solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.