Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-benzylaniline (N-phenylbenzylamine) | 103-32-2 | MFCD00003018 | 99.9% | 183.25 g/mol | C13H13N | 25 G
SDP

Catalog No. 5000303795
Encompass_Preferred

N-Benzylaniline (N-phenylbenzylamine) is an N-alkylated derivative of aniline commonly used as a research chemical and synthetic intermediate in organic and medicinal chemistry. It is supplied as a high-purity material for laboratory research use only and is not intended for human consumption.

  • High reported purity suitable for research applications.
  • Molecular formula C13H13N; molecular weight 183.25 g/mol.
  • Available in multiple pack sizes, including a 25 g solid bottle.
  • Soluble in DMSO and offered as a ready-to-use DMSO solution.
  • Typical uses include synthetic transformations and analytical reference standards.

Catalog No. 50-003-03795 Supplier Medchemexpress LLC Supplier No. 25GHYW016321
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Benzylaniline (N-phenylbenzylamine) is an N-alkylated derivative of aniline commonly used as a research chemical and synthetic intermediate in organic and medicinal chemistry. It is supplied as a high-purity material for laboratory research use only and is not intended for human consumption.

  • High reported purity suitable for research applications.
  • Molecular formula C13H13N; molecular weight 183.25 g/mol.
  • Available in multiple pack sizes, including a 25 g solid bottle.
  • Soluble in DMSO and offered as a ready-to-use DMSO solution.
  • Typical uses include synthetic transformations and analytical reference standards.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.