Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylformamide, 99%

Catalog No. AAB2372214
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2372214 25 g
AAB2372206 5 g
AAB2372222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2372214 Supplier Thermo Scientific Chemicals Supplier No. B2372214
Only null left
Add to Cart

CAS: 6343-54-0 | C8H9NO | 135.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6343-54-0
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD00003281
InChI Key IIBOGKHTXBPGEI-UHFFFAOYSA-N
Synonym benzyl formamide, n-phenylmethyl formamide, n-benzyl-formamide, unii-l363d92elk, formamide, n-phenylmethyl, formamide,n-phenylmethyl, chembl46293, n-benzyl formamide, n-benzylcarboxamide, n-formylbenzylamine
PubChem CID 80654
ChEBI CHEBI:41117
IUPAC Name N-benzylformamide
SMILES C1=CC=C(C=C1)CNC=O

Specifications

Melting Point 60°C to 61°C
Quantity 25 g
Formula Weight 135.17
Percent Purity 99%
Chemical Name or Material N-Benzylformamide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-739-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.