Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylglycine Ethyl Ester 96.0+%, TCI America™
SDP

Catalog No. B22735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22735G 5 g
B227325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22735G Supplier TCI America Supplier No. B22735G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6436-90-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.25
MDL Number MFCD00009174
InChI Key ULOLIZHBYWAICY-UHFFFAOYSA-N
Synonym n-benzylglycine ethyl ester, ethyl 2-benzylamino acetate, ethyl n-benzylglycinate, n-bzl-gly-oet, bzl-gly-oet, glycine, n-phenylmethyl-, ethyl ester, n-benzyl glycine ethyl ester, ethyl n-phenylmethyl glycinate, ethyl benzylamino acetate, n-benzyl-glycine ethyl ester
PubChem CID 80908
IUPAC Name ethyl 2-(benzylamino)acetate
SMILES CCOC(=O)CNCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 142°C
Quantity 5 g
Formula Weight 193.25
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Benzylglycine Ethyl Ester
EINECSNumber (3)-1723
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.