Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzylglycine hydrochloride, 98+%

Catalog No. AAL1520022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1520022 100 g
AAL1520014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1520022 Supplier Thermo Scientific Chemicals Supplier No. L1520022
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7689-50-1 | C9H12ClNO2 | 201.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7689-50-1
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00156934
InChI Key BUZJPENZWLUHJD-UHFFFAOYSA-N
Synonym n-benzylglycine hydrochloride, benzylaminoacetic acid hydrochloride, 2-benzylamino acetic acid hydrochloride, bzl-gly-oh.hcl, n-benzylglycine hcl, bz-gly-oh.hcl, n-benzyl glycine hydrochloride, bzl-gly-oh inverted exclamation mark currencyhcl, benzylamino acetic acid hydrochloride, glycine, n-phenylmethyl-, hydrochloride
PubChem CID 2756624
IUPAC Name 2-(benzylamino)acetic acid;hydrochloride
SMILES C1=CC=C(C=C1)CNCC(=O)O.Cl

Specifications

Melting Point ∽220°C (decomposition)
Beilstein 3915652
Quantity 100 g
Formula Weight 201.65
Percent Purity ≥98%
Chemical Name or Material N-Benzylglycine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.