Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Benzylhydroxylamine hydrochloride | 29601-98-7 | 159.62 | 100G
SDP

Catalog No. 5000447360
Encompass_Preferred

N-Benzylhydroxylamine hydrochloride is a chemical compound with the CAS number 29601-98-7 and a molecular weight of 159.62. It appears as a white to light yellow solid. This product has been tested and complies with the given specifications, showing a purity of 98.0% by NMR.

  • CAS number: 29601-98-7
  • Molecular formula: C7H10ClNO
  • Molecular weight: 159.62
  • Appearance: White to light yellow solid
  • Purity (NMR): 98.0%
  • Solid storage: 4°C, sealed, away from moisture
  • In solvent storage: -80°C for 6 months; -20°C for 1 month, sealed, away from moisture

Catalog No. 50-004-47360 Supplier Medchemexpress LLC Supplier No. HYW002216100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Benzylhydroxylamine hydrochloride is a chemical compound with the CAS number 29601-98-7 and a molecular weight of 159.62. It appears as a white to light yellow solid. This product has been tested and complies with the given specifications, showing a purity of 98.0% by NMR.

  • CAS number: 29601-98-7
  • Molecular formula: C7H10ClNO
  • Molecular weight: 159.62
  • Appearance: White to light yellow solid
  • Purity (NMR): 98.0%
  • Solid storage: 4°C, sealed, away from moisture
  • In solvent storage: -80°C for 6 months; -20°C for 1 month, sealed, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.