Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Benzylhydroxylamine hydrochloride | 29601-98-7 | MFCD00043377 | 159.62 | 500 G
SDP

Catalog No. 5000447526
Encompass_Preferred

N-Benzylhydroxylamine hydrochloride is a biochemical reagent with a purity of 98.0% (NMR). This compound, identified by CAS number 29601-98-7, appears as a white to light yellow solid. It is recommended for research use only.

  • Molecular weight: 159.62
  • Molecular formula: C7H10ClNO
  • Purity: 98.0%
  • Appearance: White to light yellow solid
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)
  • Known hazard: Irritant

Catalog No. 50-004-47526 Supplier Medchemexpress LLC Supplier No. HYW002216500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Benzylhydroxylamine hydrochloride is a biochemical reagent with a purity of 98.0% (NMR). This compound, identified by CAS number 29601-98-7, appears as a white to light yellow solid. It is recommended for research use only.

  • Molecular weight: 159.62
  • Molecular formula: C7H10ClNO
  • Purity: 98.0%
  • Appearance: White to light yellow solid
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)
  • Known hazard: Irritant

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.