Learn More
Medchemexpress LLC N-benzylideneaniline | 538-51-2 | MFCD00003027 | 100.0% | 181.23 g·mol⁻¹ | C13H11N | 10 G
SDP
Catalog No. 5000300127
Encompass_Preferred
N-benzylideneaniline is a small organic compound used as a research reagent and reported to inhibit LSD (lysine-specific demethylase). It is supplied as a solid and is characterized by molecular formula C13H11N and molecular weight 181.23 g·mol⁻¹.
  • Inhibits LSD histone demethylase for epigenetic research.
  • Supplied as a solid reagent suitable for laboratory use.
  • High purity with RP-HPLC documentation available.
  • Molecular formula C13H11N and molecular weight 181.23 g·mol⁻¹.
  • Provided with safety and technical datasheets for handling and use.
Catalog No. 50-003-00127 Supplier Medchemexpress LLC Supplier No. HYW02579910G
N-benzylideneaniline is a small organic compound used as a research reagent and reported to inhibit LSD (lysine-specific demethylase). It is supplied as a solid and is characterized by molecular formula C13H11N and molecular weight 181.23 g·mol⁻¹.
  • Inhibits LSD histone demethylase for epigenetic research.
  • Supplied as a solid reagent suitable for laboratory use.
  • High purity with RP-HPLC documentation available.
  • Molecular formula C13H11N and molecular weight 181.23 g·mol⁻¹.
  • Provided with safety and technical datasheets for handling and use.