Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyliminodiacetic Acid 98.0+%, TCI America™
SDP

Catalog No. B13635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13635G 5 g
B136325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13635G Supplier TCI America Supplier No. B13635G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3987-53-9
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.228
MDL Number MFCD00004285
InChI Key SZQUPQVVCLFZLC-UHFFFAOYSA-N
PubChem CID 77606
IUPAC Name 2-[benzyl(carboxymethyl)amino]acetic acid
SMILES C1=CC=C(C=C1)CN(CC(=O)O)CC(=O)O

Specifications

Melting Point 201°C
Color White
Quantity 5 g
Formula Weight 223.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzyliminodiacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.