Learn More
Medchemexpress LLC N-Benzyllinolenamide | 883715-18-2 | 5 MG
SDP
Catalog No. 5000421525
Encompass_Preferred
N-Benzyllinolenamide is a natural macamide isolated from Lepidium meyenii. It functions as an inhibitor of fatty acid amide hydrolase (FAAH) with an IC50 of 41.8 μM. It is for research use only and not intended for sale to patients. The product is an off-white to light yellow solid. It is also an alkaloid from plants.
  • Natural macamide isolated from Lepidium meyenii.
  • Functions as an inhibitor of fatty acid amide hydrolase (FAAH).
  • For research use only.
  • Off-white to light yellow solid.
  • Alkaloid from plants.
Catalog No. 50-004-21525 Supplier Medchemexpress LLC Supplier No. HYN30335MG
N-Benzyllinolenamide is a natural macamide isolated from Lepidium meyenii. It functions as an inhibitor of fatty acid amide hydrolase (FAAH) with an IC50 of 41.8 μM. It is for research use only and not intended for sale to patients. The product is an off-white to light yellow solid. It is also an alkaloid from plants.
  • Natural macamide isolated from Lepidium meyenii.
  • Functions as an inhibitor of fatty acid amide hydrolase (FAAH).
  • For research use only.
  • Off-white to light yellow solid.
  • Alkaloid from plants.